Learn More
Medchemexpress LLC HY-N0177 100mg Medchemexpress, Diosgenin CAS:512-04-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN0177100MG
Encompass_Preferred
Medchemexpress, HY-N0177 100mg Diosgenin CAS:512-04-9 Diosgenin, a steroidal saponin, can inhibit STAT3 signaling pathway. Diosgenin is an exogenous activator of Pdia3/ERp57. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0231