Learn More
Medchemexpress LLC Dioxopromethazine hydrochloride | 15374-15-9 | 352.88 g/mol | C17H21ClN2O2S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY107787A10MG
Encompass_Preferred
Dioxopromethazine hydrochloride is an orally active antihistamine research reagent reported to inhibit asthmatic symptoms. It is provided for laboratory research and analytical applications and is not intended for human or clinical use. Documentation available includes a product data sheet and a safety data sheet.
  • Orally active antihistamine with reported anti-asthmatic effects.
  • Intended for research and analytical applications only.
  • Available in small-scale milligram quantities suitable for screening.
  • Provided with a product data sheet and safety data sheet.
  • Molecular formula C17H21ClN2O2S, molecular weight 352.88 g/mol.
Catalog No. 50-002-40319