Learn More
Medchemexpress LLC Dioxybenzone-d3 | 131-53-3 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0966S1MG
Encompass_Preferred
Dioxybenzone-d3 is deuterium labeled Dioxybenzone. Dioxybenzone (Benzophenone-8; UV-24) is an organic compound used to block UVB and short-wave UVA (ultraviolet) radiation. It has orally activity, exhibits estrogenic disrupting effects, and up-regulates inflammatory cytokines. This compound can be used for cancer, reproductive system, and inflammatory disease studies.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes incorporated into drug molecules for quantitation during drug development.
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-08931