Learn More
Jorgensen Labs JorVet Dip Quick Stain, Introductory Kit
SDP
Supplier:  Jorgensen Labs J0322
Encompass_Preferred
For Veterinary Use Only. JorVet Dip Quick Stain is excellent for use in blood cell differential counts or as a general cytology stain, and gives coloration similar to the standard Wright-Giemsa stain. The rapid 3-step procedure can be done right in the wide-mouth 180ml. container in 15 seconds. Introductory Kit. Includes one wide-mouth bottle (180ml) of each of the 3 different components.
Catalog No. NC9581034