Learn More
Medchemexpress LLC Dipentyl phthalate-3,4,5,6-d4 | 358730-89-9 | 99.3% | 310.42 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW013816S10MG
Encompass_Preferred
Dipentyl phthalate-3,4,5,6-d4 is the deuterium labeled Dipentyl phthalate-3,4,5,6. This compound is intended for research use only and is not for sale to patients.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-54487