Learn More
Apexbio Technology LLC Diphemanil Methylsulfate 62-97-5 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A84145.1
Encompass_Preferred
Diphemanil Methylsulfate (CAS 62-97-5) is a synthetic quaternary ammonium compound that acts as an anticholinergic agent by competitively inhibiting muscarinic acetylcholine receptors This blockade reduces the activity of the parasympathetic nervous system leading to decreased secretory and smooth muscle activity Diphemanil Methylsulfate is utilized in biomedical research to investigate cholinergic pathways autonomic nervous system modulation and the physiological regulation of glandular secretions or gastrointestinal motility Its defined mechanism makes it a useful tool in studies of receptor pharmacology and neurotransmitter-mediated signaling
Catalog No. 50-000-94472