Learn More
Medchemexpress LLC Diphenhydramine | 58-73-1 | 99.6% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB030310MM/1ML
Encompass_Preferred
Diphenhydramine is a first-generation histamine H1-receptor antagonist that also exhibits anti-cholinergic effects. This compound is known for its ability to cross the ovine blood-brain barrier (BBB) and can block NMDA-activated membrane currents, which may contribute to its sedative, analgesic, and memory-related effects.
  • First-generation histamine H1-receptor antagonist
  • Possesses anti-cholinergic effect
  • Can cross the ovine blood-brain barrier
  • Targets H1 receptor and NMDA receptor
  • Blocks NMDA-activated membrane currents
  • May improve kidney injury induced by cisplatin in mice
  • Exhibits better oral bioavailability when combined with dimenhydrinate
Catalog No. 50-004-06448