Learn More
Medchemexpress LLC Diphenhydramine | 58-73-1 | 99.6% | 250 MG

Supplier: Medchemexpress LLC 250MGHYB0303
Diphenhydramine is a first-generation histamine H1-receptor antagonist with an anti-cholinergic effect. Diphenhydramine hydrochloride can cross the ovine blood-brain barrier (BBB).
- Histamine H1-receptor antagonist
- Anti-cholinergic effect
- Can cross the ovine blood-brain barrier (BBB)
- Molecular weight: 255.35
- Molecular formula: C17H21NO
- Liquid appearance
- Colorless to light yellow
- In vitro solubility in DMSO: 100 mg/mL
- Targets H1 receptor and NMDA receptor
- IC50 for NMDA receptor: 24.6 μM
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.