Learn More
Medchemexpress LLC Diphenhydramine-d6 hydrochloride | 1189986-72-8 | 99.3% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0303AS1MG
Encompass_Preferred
Diphenhydramine-d6 hydrochloride is a deuterium labeled form of Diphenhydramine hydrochloride. It functions as a first-generation histamine H1-receptor antagonist with anti-cholinergic effects, capable of crossing the ovine blood-brain barrier (BBB). This compound is valuable as a tracer and an internal standard for quantitative analysis in techniques such as NMR, GC-MS, or LC-MS. Deuterium labeling also offers potential for studying and influencing drug pharmacokinetic and metabolic profiles.
  • Deuterium labeled compound
  • First-generation histamine H1-receptor antagonist
  • Exhibits anti-cholinergic effect
  • Crosses the ovine blood-brain barrier (BBB)
  • Useful as a tracer in drug development
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-06354