Learn More
Medchemexpress LLC Diphenhydramine hydrochloride | 147-24-0 | 99.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0303A10MM/1ML
Encompass_Preferred
Diphenhydramine hydrochloride is a first-generation histamine H1-receptor antagonist with anti-cholinergic effects, capable of crossing the ovine blood-brain barrier.
  • Acts as a first-generation histamine H1-receptor antagonist
  • Exhibits anti-cholinergic effects
  • Can cross the ovine blood-brain barrier
  • Inhibits cisplatin-induced cell death in kidney cells
  • Induces apoptosis in certain cell lines
  • Reduces mortality in rats exposed to dichlorvos
Catalog No. 50-004-06073