Learn More
Medchemexpress LLC Diphenhydramine hydrochloride | 147-24-0 | 99.9% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0303AR100MG
Encompass_Preferred
Diphenhydramine hydrochloride (Standard) is an analytical standard used for research and analytical applications. It is a first-generation histamine H1-receptor antagonist with anticholinergic effects and can cross the ovine blood-brain barrier (BBB). This standard is commonly employed in qualitative, quantitative, and methodological research experiments.
  • First-generation histamine H1-receptor antagonist
  • Can cross the ovine blood-brain barrier (BBB)
  • Suitable for HPLC, GC, and MS research experiments
  • Used in qualitative, quantitative, and methodological research
Catalog No. 50-004-06019