Learn More
Medchemexpress LLC Diphenhydramine hydrochloride | 147-24-0 | 99.9% | 5 G

Supplier: Medchemexpress LLC HYB0303A5G
Diphenhydramine hydrochloride is a first-generation histamine H1-receptor antagonist with an anti-cholinergic effect, capable of crossing the ovine blood-brain barrier (BBB). It is intended for research use only.
- First-generation histamine H1-receptor antagonist
- Anti-cholinergic effect
- Capable of crossing the ovine blood-brain barrier (BBB)
- 99.93% purity
- Molecular weight: 291.82
- Formula: C17H22ClNO
- Appearance: White to off-white solid
- Inhibits Cisplatin-induced cell death in kidney proximal tubular cells
- Induces apoptosis in CCRF-CEM and Jurkat cells
- Reduces mortality in rats with acute, severe dichlorvos exposure
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.