Learn More
Medchemexpress LLC Diphenhydramine hydrochloride | 147-24-0 | 99.93% | 25 G

Supplier: Medchemexpress LLC 25GHYB0303A
Diphenhydramine hydrochloride is a first-generation histamine H1-receptor antagonist with anti-cholinergic effect. This white to off-white solid has a molecular formula of C17H22ClNO and a molecular weight of 291.82. It has been shown to cross the ovine blood-brain barrier (BBB) and is suitable for laboratory chemical applications.
- First-generation histamine H1-receptor antagonist
- Anti-cholinergic effect
- Can cross the ovine blood-brain barrier (BBB)
- Purity by LCMS: 99.93%
- Appearance: White to off-white solid
- Molecular formula: C17H22ClNO
- Molecular weight: 291.82
- Recommended storage: 4°C, sealed, away from moisture and light
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.