Learn More
U.S. Pharmacopeia Diphenhydramine Citrate, 88637-37-0, MFCD08061593, 200mg
Supplier:  U.S. Pharmacopeia 1217909
Encompass
Molecular formula C17H21NO.C6H8O7, Molecular weight 447.48, Melting Point 294.8 - 302 °F (146 - 150 °C), Synonyms: Benzhydramine citrate
Catalog No. NC0227157