Learn More
Cayman Chemical DIphenhydramInhydroCLOrId250mg
SDP
Supplier:  Cayman Chemical 11158250 MG
Encompass_Preferred
A first generation antihistamine that is a potent antagonist of the histamine H1 receptor (Ki = 11.7 nM using human recombinant receptors); readily crosses the blood-brain barrier and produces diverse cognitive and psychomotor effects; also antagonizes muscarinic cholinergic receptors (Ki = 100 to 260 nM for M1-M5)
Catalog No. 50-295-1193