Learn More
Cayman Chemical DIphenIdInhydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 1551810 MG
Encompass_Preferred
An analytical reference standard standard categorized as a dissociative anesthetic; has been found in samples seized by law enforcement; has a high potential for abuse; intended for research and forensic applications
Catalog No. 50-297-2300