Learn More
Medchemexpress LLC Diphenyl Phosphate | 838-85-7 | 250.19 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0081515G
Encompass_Preferred
Diphenyl Phosphate is a metabolic inhibitor that impacts growth and energy metabolism in zebrafish and mice in a sex-specific manner. It functions by inhibiting the activity of Succinate Dehydrogenase (SDH), which is a component of respiratory complex II. Additionally, it reduces the expression of Carnitine Palmitoyltransferase 1 (CPT1) and disrupts the integrity of the mitochondrial membrane. This compound is suitable for research related to metabolic diseases.
  • Inhibits growth and energy metabolism.
  • Specifically affects SDH (respiratory complex II) and CPT1.
  • Disrupts mitochondrial membrane integrity.
  • Useful in research on metabolic diseases.
Catalog No. 50-004-46184