Learn More
Medchemexpress LLC Diphenyl phosphate | 838-85-7 | MFCD00003033 | 250.19 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW008151
Encompass_Preferred
Diphenyl Phosphate inhibits growth and energy metabolism in zebrafish and mice in a sex-specific manner. It can inhibit the activity of SDH (respiratory complex II), reduce the expression of CPT1, and disrupt the integrity of the mitochondrial membrane. It may be used in research on metabolic diseases.
  • Inhibits growth and energy metabolism in zebrafish and mice in a sex-specific manner
  • Inhibits the activity of SDH (respiratory complex II)
  • Reduces the expression of CPT1
  • Disrupts the integrity of the mitochondrial membrane
  • May be used in research on metabolic diseases
  • Targets Succinate Dehydrogenase (SDH)
  • Targets Carnitine Palmitoyltransferase 1 (CPT1)
Catalog No. 50-004-48148