Learn More
Medchemexpress LLC Diphenylacetic acid | 117-34-0 | MFCD00004251 | 99.8% | 212.25 g/mol | C14H12O2 | 50g
SDP
Supplier:  Medchemexpress LLC HYY054950G
Encompass_Preferred
Diphenylacetic acid (Diphenylmethane- -carboxylic Acid) is a biochemical reagent that can be used as a biological material or organic compound for life science related research Diphenylacetic acid can be used as a reagent in the kinetic resolution of racemic 2-hydroxy-y-butyrolactones by asymmetric esterification in the presence of pivalic anhydride and chiral acyl-transfer catalyst Diphenylacetic acid can act as a catalyst to synthesize 2-allyl-3-oxazolin-5-one derivatives via Rh-catalyzed coupling reaction of azlactones and alkynes followed by aza-Cope rearrangement[1][2][3][4]
Catalog No. 50-002-69279