Learn More
Strem, An Ascensus Company CAS# 80-10-4. 250g. Diphenyldichlorosilane, min. 97%. MFCD00000489
SDP
Supplier:  Strem, An Ascensus Company 931407250G
Encompass
CAS# 80-10-4. 250g. Diphenyldichlorosilane, min. 97%. MFCD00000489. Molecular Weight: 253.22. Molecular Formula: (C6H5)2SiCl2. Color/form: colorless liq. Strem# 93-1407. http://www.strem.com/catalog/v/93-1407/
Catalog No. 50-901-14088
 Restricted Item