Learn More
Medchemexpress LLC Diphenylmethane (Benzylphenyl) | 101-81-5 | 99.65% | 168.24 | 500 G
SDP
Supplier:  Medchemexpress LLC HYW008814500G
Encompass_Preferred
Diphenylmethane is an aromatic compound utilized as an intermediate in drug synthesis. It finds application in the fragrance industry as an oxidizing agent and in soaps, acts as a synergist in pesticides, and serves as a plasticizer in dyes.
  • Used in drug synthesis.
  • Functions as an oxidizing agent in fragrances and soaps.
  • Acts as a synergist in pesticides.
  • Used as a plasticizer in dyes.
Catalog No. 50-004-52648