Learn More
Medchemexpress LLC Diphenylmethanimine | 1013-88-3 | 181.23 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW00761610MM/1ML
Encompass_Preferred
Diphenylmethanimine is a biochemical reagent primarily used as a biological material or organic compound for life science research. It plays a role in the preparation of nitrogen ylide dimers and functions as an ammonia surrogate. This compound also serves as an intermediate for the synthesis of various organic chemicals. Notably, it facilitates the catalytic enantioselective alkylation of glycine t-butyl ester's benzophenone imine, yielding either enantiomer of amino acid derivatives with high efficiency.
  • Used in the preparation of nitrogen ylide dimers
  • Serves as an ammonia surrogate
  • Intermediate for organic chemical synthesis
  • Facilitates catalytic enantioselective alkylation for amino acid derivatives
Catalog No. 50-004-45970