Learn More
Medchemexpress LLC Diphenylmethanimine | 1013-88-3 | 181.23 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW007616100G
Encompass_Preferred
Diphenylmethanimine is a biochemical reagent used in life science research as a biological material or organic compound.
  • It is useful in the preparation of nitrogen ylide dimers.
  • It functions as an ammonia surrogate in certain reactions.
  • It serves as an intermediate material for the synthesis of various organic chemicals.
  • It can be used in catalytic enantioselective alkylation of the benzophenone imine of glycine t-butyl ester, yielding amino acid derivatives with excellent yields when using phosphazene (Schwesinger) bases and chiral quaternary ammonium salts.
Catalog No. 50-004-45755