Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Diphenylmethanimine | 1013-88-3 | 181.23 | 250 G
SDP

Supplier:  Medchemexpress LLC 250GHYW007616

Encompass_Preferred

Diphenylmethanimine is a biochemical reagent used as a biological material or organic compound for life science-related research. It is useful in the preparation of nitrogen ylide dimers and can serve as an ammonia surrogate. This compound also functions as an intermediate material for the synthesis of organic chemicals. Additionally, it is employed in the catalytic enantioselective alkylation of the benzophenone imine of glycine t-butyl ester, utilizing phosphazene (Schwesinger) bases and chiral quaternary ammonium salts to produce amino acid derivatives in excellent yield.

  • Appearance: Liquid (density: 1.08 g/cm³)
  • Color: Colorless to light yellow
  • Molecular formula: C13H11N
  • SMILES: N=C(C1=CC=CC=C1)C2=CC=CC=C2
  • Solubility (in vitro): ≥ 100 mg/mL (551.79 mM) in DMSO

Catalog No. 50-004-50033


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.