Learn More
Medchemexpress LLC Diphenylmethanimine | 1013-88-3 | MFCD00001760 | 181.23 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW00761650G
Encompass_Preferred
Diphenylmethanimine is a biochemical reagent useful in life science research. It can be used in the preparation of nitrogen ylide dimers, as an ammonia surrogate, and as an intermediate for organic chemical synthesis. It also enables catalytic enantioselective alkylation for amino acid derivative synthesis.
  • Versatile biochemical reagent for life science applications
  • Enables the preparation of nitrogen ylide dimers
  • Functions as an effective ammonia surrogate
  • Valuable intermediate for synthesizing organic chemicals
  • Supports catalytic enantioselective alkylation to yield amino acid derivatives
  • Store in a well-ventilated place with a tightly closed container
  • Keep product stored in a locked area
  • Consult product label for specific storage temperature recommendations
Catalog No. 50-004-47551