Learn More
Medchemexpress LLC Diphenylmethylene-glycine benzyl ester | 81477-91-0 | MFCD08062206 | 98.0% | 329.40 | C22H19NO2 | 25 G

Supplier: Medchemexpress LLC 25GHYW018620
Diphenylmethylene-glycine benzyl ester is an N-protected glycine derivative used as a synthetic intermediate in peptide and small-molecule synthesis. It serves as a diphenylmethylene (DPM) protected glycine benzyl ester to facilitate selective amine protection and deprotection during multi-step syntheses.
- Provides an N-protected glycine benzyl ester useful as a synthetic intermediate.
- Supplied as a white to off-white solid powder.
- High purity suitable for synthesis (98.0%).
- Molecular formula C22H19NO2 and molecular weight 329.40.
- Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
- Available in common lab pack sizes including 25 g and 100 g.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.