Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Diphenylsulfane-d1 | 180802-01-1 | 99.5% | 196.33 g/mol | C12D10S | 10 MG
SDP

Supplier:  Medchemexpress LLC HYW016174S10MG

Encompass_Preferred

Diphenylsulfane-d1 is the deuterium-labeled analogue of diphenylsulfane used as an isotopic standard or research reagent for analytical and synthetic applications. It is supplied as a high-purity liquid and should be handled and stored according to safety data sheet recommendations to avoid exposure risks.

  • Deuterium-labeled compound with formula C12D10S.
  • High purity: 99.5% (GC).
  • Molecular weight 196.33 g/mol.
  • Colorless to light yellow liquid appearance.
  • Suitable as an isotopic standard and synthetic intermediate.
  • Recommended storage: pure form -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Hazard considerations include potential oral toxicity, skin and eye irritation, and respiratory tract irritation.

Catalog No. 50-003-02222


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.