Learn More
Cayman Chemical DIphenylterazIn 10mg
SDP
Supplier:  Cayman Chemical 3733410 MG
Encompass_Preferred
A synthetic bioluminescent luciferin; em = 502 nm; has been used to quantify extracellular hydrogen peroxide when coupled to a boronated ester cage
Catalog No. 50-291-9301