Learn More
Medchemexpress LLC HY-111382 1mg , Diphenylterazine CAS:344940-63-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1113821MG
Encompass_Preferred
HY-111382 1mg Diphenylterazine CAS: 344940-63-2 Diphenylterazine (DTZ) is a bioluminescence agent. Diphenylterazine alone yielded very little background, leading to excellent signal-to-background ratios. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9310