Learn More
Medchemexpress LLC Dipivaloylmethane | 1118-71-4 | 99.4% | 184.28 | 10 G
SDP
Supplier:  Medchemexpress LLC HY3441710G
Encompass_Preferred
Dipivaloylmethane is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • Used in the synthesis of α-aryl-β-diketones.
  • Used in the synthesis of dicyanamidobenzene-bridge diruthenium complex.
Catalog No. 50-004-04456