Learn More
Medchemexpress LLC Dipivaloylmethane | 1118-71-4 | 99.4% | 184.28 | 100 G
SDP
Supplier:  Medchemexpress LLC HY34417100G
Encompass_Preferred
Dipivaloylmethane is a biochemical reagent suitable for life science-related research. It functions as a biological material or organic compound and is used in various syntheses. This product is intended for research use only and is not sold to patients.
  • Can be used as a biological material or organic compound for life science research.
  • Used in the synthesis of α-aryl-β-diketones.
  • Used in the synthesis of dicyanamidobenzene-bridge diruthenium complex.
  • Intended for research use only.
Catalog No. 50-004-04460