Learn More
Medchemexpress LLC Dipivefrin hydrochloride | 64019-93-8 | 98.4% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB13235MG
Encompass_Preferred
Dipivefrin hydrochloride is an antiglaucoma proagent that is hydrolyzed by esterases in the cornea to produce the active compound, epinephrine. It is intended for research use only and not for human use or sale to patients.
  • Antiglaucoma proagent.
  • Hydrolyzed to active compound, epinephrine, by esterases in the cornea.
Catalog No. 50-004-11566