Learn More
Medchemexpress LLC Dipivefrin hydrochloride | 64019-93-8 | 98.40% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB132350MG
Encompass_Preferred
Dipivefrin hydrochloride is an antiglaucoma proagent that is hydrolyzed to the active compound, epinephrine, by esterases in the cornea. This compound induces enhancement of memory involving central beta- but not alpha-adrenergic mechanisms. It is intended for research use only.
  • Acts as an antiglaucoma proagent
  • Hydrolyzed to epinephrine in the cornea
  • Enhances memory
  • Involves central beta-adrenergic mechanisms
  • Suitable for in vitro and in vivo studies
  • Provided as a solid form
  • Soluble in DMSO and water
Catalog No. 50-004-12078