Learn More
Medchemexpress LLC Dipquo | 1269365-82-3 | 98.7% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY12859110MG
Encompass_Preferred
DIPQUO is an activator of the bone marker alkaline phosphatase (ALP), with an EC50 of 6.27 μM in C2C12 cells. It promotes mouse and human osteoblast differentiation through the activation of p38 MAPK-β.
  • Activator of bone marker alkaline phosphatase (ALP)
  • EC50 of 6.27 μM in C2C12 cells
  • Promotes mouse and human osteoblast differentiation
  • Activates p38 MAPK-β
Catalog No. 50-003-41893