Learn More
Medchemexpress LLC DIPQUO | 1269365-82-3 | 98.7% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY128591100MG
Encompass_Preferred
DIPQUO is an activator of the bone marker alkaline phosphatase (ALP), with an EC50 of 6.27 μM in C2C12 cells. It promotes mouse and human osteoblast differentiation through the activation of p38 MAPK-β.
  • Activates alkaline phosphatase (ALP)
  • EC50 of 6.27 μM in C2C12 cells
  • Promotes osteoblast differentiation
  • Activates p38 MAPK-β
Catalog No. 50-003-41894