Learn More
Medchemexpress LLC DIPQUO | 1269365-82-3 | >98.7% | 50 MG

Supplier: Medchemexpress LLC HY12859150MG
DIPQUO is an activator of the bone marker alkaline phosphatase (ALP), with an EC50 of 6.27 μM in C2C12 cells. It promotes mouse and human osteoblast differentiation via activation of p38 MAPK-β.
- Activates alkaline phosphatase (ALP) with an EC50 of 6.27 μM in C2C12 cells.
- Promotes mouse and human osteoblast differentiation.
- Molecular weight of 315.37.
- Chemical formula: C20H17N3O.
- Appears as a light yellow to yellow solid.
- Powder storage: -20°C for 3 years, 4°C for 2 years.
- Storage in solvent: -80°C for 6 months, -20°C for 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.