Learn More
Medchemexpress LLC Dipsanoside A | 889678-62-0 | 5 MG

Supplier: Medchemexpress LLC HYN22385MG
Dipsanoside A is a novel tetrairidoid glucoside extracted from *Dipsacus asper*. This perennial plant has a history of use in traditional Chinese medicine for its tonic properties, promotion of osseous cell fissiparism, and as an embryo security agent.
- Molecular formula: C66H90O37
- Molecular weight: 1475.40
- Appearance: Solid, white to off-white
- Isolated from Dipsacus asper
- Store at 4°C, sealed, away from moisture
- In solvent, store at -80°C for 6 months or -20°C for 1 month
- Available documentation includes data sheet, SDS, and COA
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.