Learn More
Cayman Chemical DIpyrIdamole 25g
SDP
Supplier:  Cayman Chemical 1818925 G
Encompass_Preferred
A PDE5A inhibitor (IC50 = 574 nM) that prevents platelet aggregation by increasing cGMP levels and blocking the reuptake of adenosine via red blood cells; scavenges the free radicals that inactivate COX, leading to the inhibition of platelet activation and thrombin generation; also inhibits PDE11A (IC50 = 370 nM) and ENT1 (Ki = 8.18 nM); inhibits replication of SARS-CoV-2 in Vero E6 cells at 100 nM
Catalog No. 50-296-5074