Learn More
Selleck Chemical LLC Dipyridamole S1895-10mM/1mL
SDP
Supplier:  Selleck Chemical LLC S189510MM/1ML
Encompass_Preferred
Dipyridamole is a phosphodiesterase inhibitor that blocks uptake and metabolism of adenosine by erythrocytes and vascular endothelial cells
Catalog No. 50-311-0796