Learn More
Apexbio Technology LLC Dipyridamole 58-32-2 500mg
SDP
Supplier:  Apexbio Technology LLC B1933500
Encompass_Preferred
Dipyridamole (CAS 58-32-2) is an orally bioactive phosphodiesterase (PDE) inhibitor that acts primarily by inhibiting the cellular uptake and metabolism of adenosine by erythrocytes and vascular endothelial cells This mechanism elevates extracellular adenosine levels subsequently modulating adenosine-dependent pathways involved in platelet function vasodilation and coagulation processes Dipyridamole exhibits antiplatelet properties and is commonly used in research related to secondary stroke prevention Additionally it has demonstrated the ability to induce apoptosis in certain tumor cell lines highlighting potential applications in oncology research
Catalog No. 50-000-61992