Learn More
Medchemexpress LLC Dipyridamole | 58-32-2 | 99.9% | 504.63 | 500 MG
SDP
Supplier:  Medchemexpress LLC 500MGHYB0312
Encompass_Preferred
Dipyridamole is an orally active phosphodiesterase (PDE) inhibitor and an antiplatelet agent. It is used in secondary prophylaxis against stroke and can induce cancer cell-specific apoptosis.
  • Orally active PDE inhibitor
  • Antiplatelet agent
  • Used in secondary prophylaxis against stroke
  • Can induce cancer cell-specific apoptosis
  • Targets phosphodiesterase and apoptosis
  • Relevant for cardiovascular disease research
  • Relevant for cancer research
Catalog No. 50-003-65915