Learn More
Medchemexpress LLC Dipyridamole | 58-32-2 | 99.93% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB031210MM/1ML
Encompass_Preferred
Dipyridamole is an orally active phosphodiesterase (PDE) inhibitor and antiplatelet agent. It is used for secondary prophylaxis against stroke and has been shown to induce cancer cell-specific apoptosis.
  • Orally active phosphodiesterase (PDE) inhibitor
  • Acts as an antiplatelet agent
  • Induces cancer cell-specific apoptosis
  • Increases intracellular cAMP levels
  • Induces apoptosis in primary AML cells when combined with statin
  • Mitigates tumor growth in mouse models
Catalog No. 50-004-06900