Learn More
Selleck Chemical LLC Dipyridamole 50mg 58-32-2 NSC-515776
SDP
Supplier:  Selleck Chemical LLC S189550MG
Encompass_Preferred
Dipyridamole (Persantine, NSC-515776) is a phosphodiesterase inhibitor that blocks uptake and metabolism of adenosine by erythrocytes and vascular endothelial cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-515-3