Learn More
Cayman Chemical DIquat-d4bromIde 1mg
SDP
Supplier:  Cayman Chemical 182501 MG
Encompass_Preferred
An internal standard for the quantification of diquat by GC- or LC-MS
Catalog No. 50-297-9744