Learn More
Medchemexpress LLC Diquat (dibromide hydrate) | 6385-62-2 | 99.92% | 362.06 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13637250MG
Encompass_Preferred
Diquat dibromide hydrate is a comprehensive herbicide that increases the production of reactive oxygen species (ROS) and triggers mitochondrial autophagy. It generates free radicals such as superoxide anions through redox cycles, which induce oxidative stress. The compound is cytotoxic, reproductive, and neurotoxic, and is utilized in crops like cotton and soybean to control noxious weeds.
  • Comprehensive herbicide for various crops
  • Increases production of reactive oxygen species (ROS)
  • Triggers mitochondrial autophagy
  • Generates free radicals and induces oxidative stress
  • Exhibits cytotoxic, reproductive, and neurotoxic properties
Catalog No. 50-003-48506