Learn More
Medchemexpress LLC Direct Black 38 | 1937-37-7 | 1 G
SDP
Supplier:  Medchemexpress LLC HYD02561G
Encompass_Preferred
Direct Black 38 (Chlorazol Black E) is a benzidine-based azo dye primarily used for textile dyeing. It is biodegradable when reduced by microbial enzymes, breaking its azo bonds to release benzidine and 4-aminobiphenyl. It can be used in environmental toxicology, wastewater treatment, and carcinogenicity studies.
  • Benzidine-based azo dye and substrate of Enterococcus gallinarum.
  • Primarily used for textile dyeing.
  • Biodegradable by microbial enzymes.
  • Suitable for environmental toxicology, wastewater treatment, and carcinogenicity studies.
  • Appears as a solid, light brown to black powder.
  • Soluble in DMSO (25 mg/mL) and H2O (5 mg/mL) with sonication.
Catalog No. 50-004-10980