Learn More
Medchemexpress LLC Direct Black 38 | 1937-37-7 | 10 G
SDP
Supplier:  Medchemexpress LLC HYD025610G
Encompass_Preferred
Direct Black 38 (Chlorazol Black E) is a benzidine-based azo dye. Primarily used for textile dyeing, it is biodegradable through reduction by microbial enzymes, which breaks its azo bonds to release benzidine and 4-aminobiphenyl. This compound is valuable in environmental toxicology, wastewater treatment, and carcinogenicity studies, though its metabolite benzidine is classified as a human carcinogen.
  • Benzidine-based azo dye
  • Primarily used for textile dyeing
  • Biodegradable by microbial enzymes
  • Relevant in environmental toxicology, wastewater treatment, and carcinogenicity studies
  • Metabolite benzidine is a human carcinogen
  • Decolorized by Enterococcus gallinarum
  • Soluble in DMSO and H2O
Catalog No. 50-004-10784