Learn More
Medchemexpress LLC Direct Blue 1 (Chicago Sky Blue 6B) | 2610-05-1 | 95.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYD093910MM/1ML
Encompass_Preferred
Direct Blue 1 (Chicago Sky Blue 6B) is a complex dye used for background autofluorescence in immunofluorescence histochemistry. It is a potent and competitive VGLUT inhibitor and can inhibit the Aβ-binding small molecule PrP ligand. It also demonstrates anti-inflammatory activity.
  • Binds α-synuclein and inhibits its aggregation.
  • Blocks proliferation of α-synuclein between SH-SY5Y cells.
  • Inhibits RANKL-induced osteoclasts and bone resorption in vitro by inhibiting NF-κB signaling.
  • Promotes proteasome-mediated MIF degradation.
  • Alleviates inflammatory bone destruction and osteoporosis induced by titanium particles in mice.
  • Attenuates methamphetamine-induced hyperactivity and behavioral sensitization in mice.
Catalog No. 50-004-09609