Learn More
Enzo Life Sciences Enzo Life Sciences Direct cAMP ELISA Kit, Quantitative cAMP Determination, Competitive Assay, 96 Well
SDP
Supplier:  Enzo Life Sciences ADI900066A
Encompass_Preferred
The Enzo Life Sciences Direct cAMP ELISA Kit is a competitive immunoassay designed for the quantitative measurement of intracellular cyclic AMP (cAMP) in various biological samples. This colorimetric kit streamlines the analysis process by allowing direct assessment of samples lysed in 0.1 M HCl, which saves time and enhances sensitivity. An optional acetylation protocol significantly increases sensitivity, making this kit ideal for low-concentration samples.
Catalog No. NC2068019