Learn More
Selleck Chemical LLC DIRECT RED 80 E2905-100MG
SDP
Supplier:  Selleck Chemical LLC E2905100MG
Encompass_Preferred
Direct Red 80 (Sirius Red) is a polyazo dye used principally in staining methods for collagen and amyloid Direct Red 80 does not release benzidine upon degradation and is safer than many traditional direct dyes
Catalog No. 50-000-40540